Thermo Scientific Chemicals

Pyruvic aldehyde, 35-45% w/w aq. soln, Thermo Scientific Chemicals

Catalog number: B24664.36
500 g, Each
Thermo Scientific Chemicals

Pyruvic aldehyde, 35-45% w/w aq. soln, Thermo Scientific Chemicals

Catalog number: B24664.36
500 g, Each
Quantity
Catalog number: B24664.36
also known as B24664-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-98-8
IUPAC Name
2-oxopropanal
Molecular Formula
C3H4O2
InChI Key
AIJULSRZWUXGPQ-UHFFFAOYSA-N
SMILES
CC(=O)C=O
Appearance (Color)
Clear colorless to yellow to orange
Acid-base back titration
28-50%
Comment
RI variable with level of polymerisation
Refractive Index
1.3910-1.4200 @ 20°C
Form
Liquid

Description

Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

Solubility
Soluble in DMSO, water.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text