Thermo Scientific Chemicals

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B24678.06
5 g, Each
Thermo Scientific Chemicals

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B24678.06
5 g, Each
Quantity
Catalog number: B24678.06
also known as B24678-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4042-36-8
IUPAC Name
5-oxopyrrolidine-2-carboxylic acid
Molecular Formula
C5H7NO3
InChI Key
ODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILES
OC(=O)C1CCC(=O)N1
Appearance (Color)
White to cream
Form
Crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
151-163?C
Optical Rotation
+10.5? ? 1? (c=5 in water)

Description

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing /reducing agents, strong acids, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text