Thermo Scientific Chemicals

4-Ethylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B24682.14
25 g, Each
Thermo Scientific Chemicals

4-Ethylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B24682.14
25 g, Each
Quantity
Catalog number: B24682.14
also known as B24682-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16712-69-9
IUPAC Name
4-ethylbenzene-1-sulfonyl chloride
Molecular Formula
C8H9ClO2S
InChI Key
LACFLXDRFOQEFZ-UHFFFAOYSA-N
SMILES
CCC1=CC=C(C=C1)S(Cl)(=O)=O
Refractive Index
1.5430-1.5480 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Reacts with water. Soluble in ethanol, acetone and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases, amines and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text