5-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
5-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24684.03
also known as B24684-03
Price (USD)/ Each
43.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
43.10
Add to cart
5-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB24684.03
Price (USD)/ Each
43.10
-
Add to cart
Chemical Identifiers
CAS19415-51-1
IUPAC Name5-fluoro-2-methoxybenzaldehyde
Molecular FormulaC8H7FO2
InChI KeyCRLDWFVRQNUUSZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(F)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Melting Point (clear melt)44.0-50.0?C
Appearance (Color)Pale yellow to yellow or cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only