Thermo Scientific Chemicals

2,6-Dimethylpyrazine, 99%, Thermo Scientific Chemicals

Catalog number: B24687.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dimethylpyrazine, 99%, Thermo Scientific Chemicals

Catalog number: B24687.14
25 g, Each
Quantity
Catalog number: B24687.14
also known as B24687-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-50-9
IUPAC Name
2,6-dimethylpyrazine
Molecular Formula
C6H8N2
InChI Key
HJFZAYHYIWGLNL-UHFFFAOYSA-N
SMILES
CC1=CN=CC(C)=N1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥98.5%
Melting Point (clear melt)
33.5-39.5?C

Description

2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text