Thermo Scientific Chemicals

Methyl diphenylacetate, 98%, Thermo Scientific Chemicals

Catalog number: B24694.22
100 g, Each
Thermo Scientific Chemicals

Methyl diphenylacetate, 98%, Thermo Scientific Chemicals

Catalog number: B24694.22
100 g, Each
Quantity
Catalog number: B24694.22
also known as B24694-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3469-00-9
IUPAC Name
methyl 2,2-diphenylacetate
Molecular Formula
C15H14O2
InChI Key
AORIUCNKPVHMTN-UHFFFAOYSA-N
SMILES
COC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
56.0-62.0?C
Appearance (Color)
White

Description

Methyl diphenylacetate is used as a chemical, organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diphenylacetate is used as a chemical, organic intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text