Thermo Scientific Chemicals

Ethyl thiophene-2-glyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: B24712.09
10 g, Each
Thermo Scientific Chemicals

Ethyl thiophene-2-glyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: B24712.09
10 g, Each
Quantity
Catalog number: B24712.09
also known as B24712-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4075-58-5
IUPAC Name
ethyl 2-oxo-2-(thiophen-2-yl)acetate
Molecular Formula
C8H8O3S
InChI Key
GHOVLEQTRNXASK-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=O)C1=CC=CS1
Refractive Index
1.5475-1.5525 @ 20?C
Assay (GC)
≥96.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow

Description

Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text