Nipecotic acid, 98%, Thermo Scientific Chemicals
Nipecotic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nipecotic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24723.03
also known as B24723-03
Price (USD)/ Each
35.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
35.20
Add to cart
Nipecotic acid, 98%, Thermo Scientific Chemicals
Catalog numberB24723.03
Price (USD)/ Each
35.20
-
Add to cart
Chemical Identifiers
CAS498-95-3
IUPAC Namepiperidine-3-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyXJLSEXAGTJCILF-UHFFFAOYSA-N
SMILESOC(=O)C1CCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
GABA reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only