Thermo Scientific Chemicals

4-Dimethylaminocinnamaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B24741.06
5 g, Each
Thermo Scientific Chemicals

4-Dimethylaminocinnamaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B24741.06
5 g, Each
Quantity
Catalog number: B24741.06
also known as B24741-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6203-18-5
IUPAC Name
(2E)-3-[4-(dimethylamino)phenyl]prop-2-enal
Molecular Formula
C11H13NO
InChI Key
RUKJCCIJLIMGEP-ONEGZZNKSA-N
SMILES
CN(C)C1=CC=C(\C=C\C=O)C=C1
Appearance (Color)
Yellow to orange
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Free acid (titration)
≤1.0%

Description

4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

Solubility
Soluble in chloroform, ethanol, dioxane.

Notes
Air & light sensitive. Store at -20°C. Store in dark. Incompatible with oxidizing agents, air, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text