1,4-Diethylpiperazine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, toluene.
Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
RUO – Research Use Only
General References:
- T. J. Curphey. Heterocyclic Diquaternary Salts. J. Am. Chem. Soc. 1965, 87 (9), 2063-2064.
- Christina L. L. Chai; Graham A. Heath; Paul B. Huleatt and Gavin A. O’Shea. The first electrochemical study of epidithiopiperazine-2,5-diones, a special class of α,α'-disulfide bridged cyclic dipeptides. J. Chem. Soc., Perkin Trans. 2. 2010, (3), 389-392.