Thermo Scientific Chemicals

1,4-Diethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B24743.30
250 g, Each
Thermo Scientific Chemicals

1,4-Diethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B24743.30
250 g, Each
Quantity
Catalog number: B24743.30
also known as B24743-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6483-50-7
IUPAC Name
1,4-diethylpiperazine-1,4-diium
Molecular Formula
C8H20N2
InChI Key
DDPRYTUJYNYJKV-UHFFFAOYSA-P
SMILES
CC[NH+]1CC[NH+](CC)CC1
Identification (FTIR)
Conforms
Refractive Index
1.4520-1.4560
Appearance (Color)
Colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

1,4-Diethylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, toluene.

Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text