Thermo Scientific Chemicals

2,2'-Thenil, 98%, Thermo Scientific Chemicals

Catalog number: B24753.03
1 g, Each
Thermo Scientific Chemicals

2,2'-Thenil, 98%, Thermo Scientific Chemicals

Catalog number: B24753.03
1 g, Each
Quantity
Catalog number: B24753.03
also known as B24753-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7333-07-5
IUPAC Name
bis(thiophen-2-yl)ethane-1,2-dione
Molecular Formula
C10H6O2S2
InChI Key
UNWKVSDABPCZMK-UHFFFAOYSA-N
SMILES
O=C(C(=O)C1=CC=CS1)C1=CC=CS1
Assay (GC)
≥97.5%
Melting Point (clear melt)
81-85?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Yellow
Identification (FTIR)
Conforms

Description

Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text