Thermo Scientific Chemicals

(+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: B24754.09
10 g, Each
Thermo Scientific Chemicals

(+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: B24754.09
10 g, Each
Quantity
Catalog number: B24754.09
also known as B24754-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17026-42-5
IUPAC Name
(2S,3S)-2,3-bis(benzoyloxy)butanedioic acid
Molecular Formula
C18H14O8
InChI Key
YONLFQNRGZXBBF-ZCWZLOQUNA-N
SMILES
OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
Assay (unspecified)
Aqueous acid-base Titration OR HPLC
Assay (HPLC)
>98.5%
Optical Rotation
+120 ? 3? (c=5 in methanol)
Appearance (Color)
White
Form
Powder

Description

For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text