Thermo Scientific Chemicals

Methyl 2,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B24757.22
100 g, Each
Thermo Scientific Chemicals

Methyl 2,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B24757.22
100 g, Each
Quantity
Catalog number: B24757.22
also known as B24757-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2150-47-2
IUPAC Name
methyl 2,4-dihydroxybenzoate
Molecular Formula
C8H8O4
InChI Key
IIFCLXHRIYTHPV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(O)C=C1O
Appearance (Color)
Pale cream
Melting Point (clear melt)
113.0-122.0?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%

Description

Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text