Thermo Scientific Chemicals

Benzhydryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24764.14
25 g, Each
Thermo Scientific Chemicals

Benzhydryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24764.14
25 g, Each
Quantity
Catalog number: B24764.14
also known as B24764-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-99-3
IUPAC Name
[chloro(phenyl)methyl]benzene
Molecular Formula
C13H11Cl
InChI Key
ZDVDCDLBOLSVGM-UHFFFAOYSA-N
SMILES
ClC(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to pale green
Assay (GC)
≥97.5%
Refractive Index
1.5930-1.5970 @ 20?C
Form
Liquid

Description

Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

Solubility
Insoluble in water.

Notes
Store in a tightly sealed container. Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text