Thermo Scientific Chemicals

Benzyl n-butyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B24769.22
100 g, Each
Thermo Scientific Chemicals

Benzyl n-butyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B24769.22
100 g, Each
Quantity
Catalog number: B24769.22
also known as B24769-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85-68-7
IUPAC Name
1-benzyl 2-butyl benzene-1,2-dicarboxylate
Molecular Formula
C19H20O4
InChI Key
IRIAEXORFWYRCZ-UHFFFAOYSA-N
SMILES
CCCCOC(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.5385-1.5425 @ 20?C

Description

Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, alkalis and strong bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text