Thermo Scientific Chemicals

1-Adamantaneacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: B24770.06
5 g, Each
Thermo Scientific Chemicals

1-Adamantaneacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: B24770.06
5 g, Each
Quantity
Catalog number: B24770.06
also known as B24770-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16269-13-9
IUPAC Name
2-(adamantan-1-yl)acetonitrile
Molecular Formula
C12H17N
InChI Key
DXQVPXCZIRQITG-UHFFFAOYSA-N
SMILES
N#CCC12CC3CC(CC(C3)C1)C2
Assay (GC)
≥96.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
71.5-77.5?C

Description

1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text