Methylurea, 98%, Thermo Scientific Chemicals
Methylurea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylurea, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number B24772.0B
also known as B24772-0B
Price (USD)/ Each
206.00
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
206.00
Add to cart
Methylurea, 98%, Thermo Scientific Chemicals
Catalog numberB24772.0B
Price (USD)/ Each
206.00
-
Add to cart
Chemical Identifiers
CAS598-50-5
IUPAC Namemethylurea
Molecular FormulaC2H6N2O
InChI KeyXGEGHDBEHXKFPX-UHFFFAOYSA-N
SMILESCNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only