Thermo Scientific Chemicals

3-Pyridyloxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24780.06
5 g, Each
Thermo Scientific Chemicals

3-Pyridyloxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24780.06
5 g, Each
Quantity
Catalog number: B24780.06
also known as B24780-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86649-57-2
Appearance (Color)
White to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.0 g/L)

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text