Thermo Scientific Chemicals

Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: B24784.06
5 g, Each
Thermo Scientific Chemicals

Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: B24784.06
5 g, Each
Quantity
Catalog number: B24784.06
also known as B24784-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34156-56-4
Appearance (Color)
White
Water Content (Karl Fischer Titration)
31.94-39.65% (5-7 waters)
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Powder

Description

Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text