Thermo Scientific Chemicals

2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: B24787.22
100 g, Each
Thermo Scientific Chemicals

2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: B24787.22
100 g, Each
Quantity
Catalog number: B24787.22
also known as B24787-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-78-8
IUPAC Name
2,5-dichlorophenol
Molecular Formula
C6H4Cl2O
InChI Key
RANCECPPZPIPNO-UHFFFAOYSA-N
SMILES
OC1=CC(Cl)=CC=C1Cl
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
53.0-60.0?C
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text