Thermo Scientific Chemicals

3,4-Dimethylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24806.06
5 g, Each
Thermo Scientific Chemicals

3,4-Dimethylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24806.06
5 g, Each
Quantity
Catalog number: B24806.06
also known as B24806-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2905-30-8
IUPAC Name
3,4-dimethylbenzene-1-sulfonyl chloride
Molecular Formula
C8H9ClO2S
InChI Key
DNMQPRPJIWTNAX-UHFFFAOYSA-N
SMILES
CC1=C(C)C=C(C=C1)S(Cl)(=O)=O
Melting Point (clear melt)
48.0-54.0?C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%

Description

3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text