Thermo Scientific Chemicals

Ethyl 3,4-dihydroxycinnamate, 97%, Thermo Scientific Chemicals

Catalog number: B24808.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 3,4-dihydroxycinnamate, 97%, Thermo Scientific Chemicals

Catalog number: B24808.06
5 g, Each
Quantity
Catalog number: B24808.06
also known as B24808-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66648-50-8
IUPAC Name
ethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular Formula
C11H12O4
InChI Key
WDKYDMULARNCIS-GQCTYLIASA-N
SMILES
CCOC(=O)C=CC1=CC=C(O)C(O)=C1
Appearance (Color)
Brown
Form
Powder
Melting Point (clear melt)
142.0-148.0?C
Assay (GC)
≥96.0%

Description

Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text