Thermo Scientific Chemicals

Dimidium bromide, 95%, Thermo Scientific Chemicals

Catalog number: B24818.03
1 g, Each
Thermo Scientific Chemicals

Dimidium bromide, 95%, Thermo Scientific Chemicals

Catalog number: B24818.03
1 g, Each
Quantity
Catalog number: B24818.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
518-67-2
IUPAC Name
3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
Molecular Formula
C20H18BrN3
InChI Key
MQOKYEROIFEEBH-UHFFFAOYSA-N
SMILES
[Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤6%
Assay (Titration ex Bromide)
≥94.0 to ≤106.0% (dry wt. basis)
Appearance (Color)
Dark red or purple
lambda-max (UV-Vis)
521-526 nm (methanol)

Description

Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text