Thermo Scientific Chemicals

1-Methoxynaphthalene, 98+%, Thermo Scientific Chemicals

Catalog number: B24825.36
500 g, Each
Thermo Scientific Chemicals

1-Methoxynaphthalene, 98+%, Thermo Scientific Chemicals

Catalog number: B24825.36
500 g, Each
Quantity
Catalog number: B24825.36
also known as B24825-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2216-69-5
IUPAC Name
1-methoxynaphthalene
Molecular Formula
C11H10O
InChI Key
NQMUGNMMFTYOHK-UHFFFAOYSA-N
SMILES
COC1=C2C=CC=CC2=CC=C1
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.6190-1.6235 @ 20?C

Description

1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.

Solubility
Immiscible with water. Miscible with ether, carbon disulfide and benzene.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text