Thermo Scientific Chemicals

(2-Bromoethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: B24833.06
5 g, Each
Thermo Scientific Chemicals

(2-Bromoethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: B24833.06
5 g, Each
Quantity
Catalog number: B24833.06
also known as B24833-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7301-93-1
IUPAC Name
(2-bromoethyl)triphenylphosphanium bromide
Molecular Formula
C20H19Br2P
InChI Key
ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
SMILES
[Br-].BrCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%

Description

(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text