Thermo Scientific Chemicals

Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B24840.22
100 g, Each
Thermo Scientific Chemicals

Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B24840.22
100 g, Each
Quantity
Catalog number: B24840.22
also known as B24840-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-62-0
IUPAC Name
iodocyclohexane
Molecular Formula
C6H11I
InChI Key
FUCOMWZKWIEKRK-UHFFFAOYSA-N
SMILES
IC1CCCCC1
Assay (GC)
≥97.5%
Refractive Index
1.5460-1.5500 @ 20°C
Appearance (Color)
Clear pale yellow to yellow to orange to pale red to pale brown
Form
Liquid

Description

Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

Solubility
Soluble in ethanol, ether, acetone.

Notes
Light sensitive. Store away from oxidizing agent, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text