Thermo Scientific Chemicals

Methyl propionylacetate, 99%, Thermo Scientific Chemicals

Catalog number: B24848.14
25 g, Each
Thermo Scientific Chemicals

Methyl propionylacetate, 99%, Thermo Scientific Chemicals

Catalog number: B24848.14
25 g, Each
Quantity
Catalog number: B24848.14
also known as B24848-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30414-53-0
IUPAC Name
methyl 3-oxopentanoate
Molecular Formula
C6H10O3
InChI Key
XJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILES
CCC(=O)CC(=O)OC
Form
Liquid
Refractive Index
1.4215-1.4245 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (Silylated GC)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text