Methyl propionylacetate, 99%, Thermo Scientific Chemicals
Methyl propionylacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl propionylacetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B24848.22
also known as B24848-22
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
Methyl propionylacetate, 99%, Thermo Scientific Chemicals
Catalog numberB24848.22
Price (USD)/ Each
166.00
-
Add to cart
Chemical Identifiers
CAS30414-53-0
IUPAC Namemethyl 3-oxopentanoate
Molecular FormulaC6H10O3
InChI KeyXJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILESCCC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4215-1.4245 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only