Thermo Scientific Chemicals

1-Pentyn-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: B24853.06
5 g, Each
Thermo Scientific Chemicals

1-Pentyn-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: B24853.06
5 g, Each
Quantity
Catalog number: B24853.06
also known as B24853-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4187-86-4
IUPAC Name
pent-1-yn-3-ol
Molecular Formula
C5H8O
InChI Key
LBSKEFWQPNVWTP-UHFFFAOYNA-N
SMILES
CCC(O)C#C
Form
Liquid
Assay (GC)
>97.5%
Refractive Index
1.4320-1.4360 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

1-Pentyn-3-ol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentyn-3-ol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text