1-Pentyn-3-ol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentyn-3-ol is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
RUO – Research Use Only
E.J. Duwell; J.W. Todd; H.C. Butzke. The mechanism of corrosion inhibition of steel by ethynylcyclohexanol in acid solution. Corrosion Science. 1964, 4 (1-4), 435-441.
Fabio Bellina; Donatella Ciucci; Piergiorgio Vergamini; Renzo Rossi. Regioselective Synthesis of Natural and Unnatural (Z)-3-(1-Alkylidene)phthalides and 3-Substituted Isocoumarins Starting from Methyl 2-Hydroxybenzoates. Tetrahedron. 2000, 56 (16), 2533-2545.