6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals
6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24857.03
also known as B24857-03
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
Add to cart
6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals
Catalog numberB24857.03
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
-
Add to cart
Chemical Identifiers
CAS1191-25-9
IUPAC Name6-hydroxyhexanoate
Molecular FormulaC6H11O3
InChI KeyIWHLYPDWHHPVAA-UHFFFAOYSA-M
SMILESOCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or liquid as melt
Assay (Silylated GC)≥94.0% (monomer + dimer)
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to white
Assay (Silylated GC)Monomer content: ≥80%
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Borzenkov, M.; Mitina, N.; Lobaz, V.; Hevus, O. Synthesis and Properties of Novel Surface Active Monomers Based on Derivatives of 4-Hydroxybutyric Acid and 6-Hydroxyhexanoic Acid. J. Surfactants Deterg. 2015, 18 (1), 133-144.
  2. Clomburg, J. M.; Blankschien, M. D.; Vick, J. E.; Chou, A.; Kim, S.; Gonzalez, R. Integrated engineering of beta-oxidation reversal and μ-oxidation pathways for the synthesis of medium chain μ-functionalized carboxylic acids. Metab. Eng. 2015, 28, 202-212.