Thermo Scientific Chemicals

L-Glutamic acid 5-methyl ester, 99%

Catalog number: B24859.06
5 g, Each
Thermo Scientific Chemicals

L-Glutamic acid 5-methyl ester, 99%

Catalog number: B24859.06
5 g, Each
Quantity
Catalog number: B24859.06
also known as B24859-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1499-55-4
IUPAC Name
2-amino-5-methoxy-5-oxopentanoic acid
Molecular Formula
C6H11NO4
InChI Key
ZGEYCCHDTIDZAE-UHFFFAOYNA-N
SMILES
COC(=O)CCC(N)C(O)=O
Appearance (Color)
White
Optical Rotation
+13.0? ? 2.0? (c=1 in water)
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text