Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals
Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24861.03
also known as B24861-03
Price (USD)/ Each
28.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
28.90
Add to cart
Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals
Catalog numberB24861.03
Price (USD)/ Each
28.90
-
Add to cart
Chemical Identifiers
CAS943-37-3
IUPAC Nameethyl 5-nitrofuran-2-carboxylate
Molecular FormulaC7H7NO5
InChI KeyJMNXLAQKIHVFIC-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(O1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)95.0-104.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only