Thermo Scientific Chemicals

Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: B24861.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 5-nitro-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: B24861.06
5 g, Each
Quantity
Catalog number: B24861.06
also known as B24861-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
943-37-3
IUPAC Name
ethyl 5-nitrofuran-2-carboxylate
Molecular Formula
C7H7NO5
InChI Key
JMNXLAQKIHVFIC-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(O1)[N+]([O-])=O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
95.0-104.0?C
Appearance (Color)
Pale cream to cream
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text