Thermo Scientific Chemicals

1-Thionaphthol, 99%, Thermo Scientific Chemicals

Catalog number: B24876.06
5 g, Each
Thermo Scientific Chemicals

1-Thionaphthol, 99%, Thermo Scientific Chemicals

Catalog number: B24876.06
5 g, Each
Quantity
Catalog number: B24876.06
also known as B24876-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
529-36-2
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%

Description

1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text