Ethyl 4-phenylpyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 4-phenylpyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-phenylpyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B24877.06
also known as B24877-06
Price (USD)/ Each
297.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
297.00
Add to cart
Ethyl 4-phenylpyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB24877.06
Price (USD)/ Each
297.00
-
Add to cart
Chemical Identifiers
CAS64276-62-6
IUPAC Nameethyl 4-phenyl-1H-pyrrole-3-carboxylate
Molecular FormulaC13H13NO2
InChI KeyVNZGRYAVCWGZCT-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CNC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)123.0-133.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only