Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24887.30
250 g, Each
Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24887.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B24887.30
also known as B24887-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-91-7
IUPAC Name
3,4,5-trihydroxybenzoic acid
Molecular Formula
C7H6O5
InChI Key
LNTHITQWFMADLM-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(O)=C(O)C(O)=C1
Appearance (Color)
White to cream or pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5 %
Identification (FTIR)
Conforms

Description

Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text