Thermo Scientific Chemicals

Diacetone-D-galactose, 95%, Thermo Scientific Chemicals

Catalog number: B24899.14
25 g, Each
Thermo Scientific Chemicals

Diacetone-D-galactose, 95%, Thermo Scientific Chemicals

Catalog number: B24899.14
25 g, Each
Quantity
Catalog number: B24899.14
also known as B24899-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4064-06-6
IUPAC Name
{4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
Molecular Formula
C12H20O6
InChI Key
POORJMIIHXHXAV-UHFFFAOYNA-N
SMILES
CC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
Appearance (Color)
Clear colorless to yellow
Form
Viscous liquid or paste
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Optical Rotation
-58? ±4? (c =3 in Chloroform)

Description

Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

Solubility
Soluble in Acetone, Chloroform and Methanol.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text