Thermo Scientific Chemicals

4-Bromo-2-chlorobenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: B24913.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-chlorobenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: B24913.14
25 g, Each
Quantity
Catalog number: B24913.14
also known as B24913-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
351003-52-6
IUPAC Name
4-bromo-2-chlorobenzene-1-sulfonyl chloride
Molecular Formula
C6H3BrCl2O2S
InChI Key
OECYEHWJPRPCOH-UHFFFAOYSA-N
SMILES
ClC1=CC(Br)=CC=C1S(Cl)(=O)=O
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Assay (GC)
≥95.0%
Melting Point (clear melt)
72.0-82.0?C
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text