3-Nitro-o-xylene, 99%, Thermo Scientific Chemicals
3-Nitro-o-xylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Nitro-o-xylene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B24917.36
also known as B24917-36
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
Add to cart
3-Nitro-o-xylene, 99%, Thermo Scientific Chemicals
Catalog numberB24917.36
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS83-41-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to green
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5425-1.5455 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only