2-Fluorobenzhydrazide, 98%, Thermo Scientific Chemicals
2-Fluorobenzhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluorobenzhydrazide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B24921.09
also known as B24921-09
Price (USD)/ Each
53.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
53.00
Add to cart
2-Fluorobenzhydrazide, 98%, Thermo Scientific Chemicals
Catalog numberB24921.09
Price (USD)/ Each
53.00
-
Add to cart
Chemical Identifiers
CAS446-24-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)68.0-75.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only