Thermo Scientific Chemicals

4,4'-Dimethyl-2,2'-bipyridine, 98%, Thermo Scientific Chemicals

Catalog number: B24936.03
1 g, Each
Thermo Scientific Chemicals

4,4'-Dimethyl-2,2'-bipyridine, 98%, Thermo Scientific Chemicals

Catalog number: B24936.03
1 g, Each
Quantity
Catalog number: B24936.03
also known as B24936-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1134-35-6
IUPAC Name
4,4'-dimethyl-2,2'-bipyridine
Molecular Formula
C12H12N2
InChI Key
NBPGPQJFYXNFKN-UHFFFAOYSA-N
SMILES
CC1=CC=NC(=C1)C1=CC(C)=CC=N1
Melting Point (clear melt)
170.0-176.0?C
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

4,4'-Dimethyl-2,2'-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethyl-2,2′-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.

Solubility
Soluble in ethanol, acetic acid, benzene, toluene. Highly soluble in water at pH < 2.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text