N-Acetylglycinamide, 97%, Thermo Scientific Chemicals
N-Acetylglycinamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetylglycinamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24940.03
also known as B24940-03
Price (USD)/ Each
52.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
52.90
Add to cart
N-Acetylglycinamide, 97%, Thermo Scientific Chemicals
Catalog numberB24940.03
Price (USD)/ Each
52.90
-
Add to cart
Chemical Identifiers
CAS2620-63-5
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)132.5-141.5?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only