Benzylurea is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzylurea is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
James B.Gerken; Scott Ross. An NMR investigation of the existence of halide and carboxylate co-solute effects on the rotational barrier about the C-N bonds in urea and thiourea. Journal of Physical Organic Chemistry. 2010, 23,(9), 806-809.
Stanley J.Cristol; Richard P.Evans; Karl L.Lockwood. Photochemical transformations. 14. Photochemical reactions of ketones with some aliphatic ureas. J. Org. Chem. 1977, 42,(14), 2378-2379.