Thermo Scientific Chemicals

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: B24951.14
25 g, Each
Thermo Scientific Chemicals

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: B24951.14
25 g, Each
Quantity
Catalog number: B24951.14
also known as B24951-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
129-64-6
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Acid-base back titration
≥95.0 to <105.0%
Melting Point
163-168°C

Description

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text