4-n-Propoxybenzoic acid, 98%, Thermo Scientific Chemicals
4-n-Propoxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Propoxybenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B24958.14
also known as B24958-14
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
4-n-Propoxybenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberB24958.14
Price (USD)/ Each
116.00
-
Add to cart
Chemical Identifiers
CAS5438-19-7
IUPAC Name4-propoxybenzoic acid
Molecular FormulaC10H12O3
InChI KeyGDFUWFOCYZZGQU-UHFFFAOYSA-N
SMILESCCCOC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)143.0-149.0?C
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only