Sodium 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Sodium 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
Catalog number B24960.0I
also known as B24960-0I
Price (USD)/ Each
367.00
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
367.00
Add to cart
Sodium 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Catalog numberB24960.0I
Price (USD)/ Each
367.00
-
Add to cart
Chemical Identifiers
CAS19766-89-3
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤4.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Fabrizio Carta; Alessio Innocenti; Rebecca A Hall; Fritz A Mühlschlegel; Andrea Scozzafava; Claudiu T Supuran. Carbonic anhydrase inhibitors. Inhibition of the β-class enzymes from the fungal pathogens Candida albicans and Cryptococcus neoformans with branched aliphatic/aromatic carboxylates and their derivatives. Bioorganic & Medicinal Chemistry Letters.2011, 21, (8), 2521-2526.
  2. John Fitt; Kapa Prasad; Oljan Repič; Thomas J. Blacklock. Sodium 2-ethylhexanoate: A mild acid scavenger useful in acylation of amines. Tetrahedron Letters.1998, 39, 6991-6992.