Thermo Scientific Chemicals

Cyclopentanone oxime, 97%, Thermo Scientific Chemicals

Catalog number: B24961.22
100 g, Each
Thermo Scientific Chemicals

Cyclopentanone oxime, 97%, Thermo Scientific Chemicals

Catalog number: B24961.22
100 g, Each
Quantity
Catalog number: B24961.22
also known as B24961-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1192-28-5
IUPAC Name
N-cyclopentylidenehydroxylamine
Molecular Formula
C5H9NO
InChI Key
YGNXYFLJZILPEK-UHFFFAOYSA-N
SMILES
ON=C1CCCC1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
52.0-61.0?C
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text