3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B24963.14
also known as B24963-14
Price (USD)/ Each
171.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
171.00
Add to cart
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
Catalog numberB24963.14
Price (USD)/ Each
171.00
-
Add to cart
Chemical Identifiers
CAS10269-01-9
IUPAC Name1-(3-bromophenyl)methanamine
Molecular FormulaC7H8BrN
InChI KeySUYJXERPRICYRX-UHFFFAOYSA-N
SMILESNCC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5850-1.5900 @ 20°C (non-U.S. specification)
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Jeremy P. Mallari; Anang A. Shelat; Aaron Kosinsk; Conor R. Caffrey; Michele Connelly; Fangyi Zhu; James H. McKerrow and R. Kiplin Guy. Structure-Guided Development of Selective TbcatB Inhibitors. J. Med. Chem. 2009, 52 (20), 6489-6493.
  2. Aldo Caiazzo; Paula M. L. Garcia; Ron Wever; Jan C. M. van Hest; Alan E. Rowan and Joost N. H. Reek. Synergy between chemo- and bio-catalysts in multi-step transformations. Organic & Biomolecular Chemistry. 2009, 14 (7), 2926-2932.